Learn More
Medchemexpress LLC Cilengitide trifluoroacetate | 199807-35-7 | MFCD04307743 | 99.9% | 702.68 | C29H41F3N8O9 | 10 MG
SDP
Catalog No. 5000288866
Encompass_Preferred
Cilengitide trifluoroacetate is the trifluoroacetate (TFA) salt of cilengitide, a cyclic RGD peptide that selectively inhibits αvβ3 and αvβ5 integrin receptors. It is provided as a high-purity research reagent for studies of integrin-mediated signaling, angiogenesis, and tumor biology.
  • Selective inhibitor of αvβ3 and αvβ5 integrins (IC50: 4 nM and 79 nM).
  • Supplied as the trifluoroacetate (TFA) salt form for research use.
  • High reported purity (~99.9%) suitable for laboratory studies.
  • Available in small research pack sizes (for example, 10 mg).
Catalog No. 50-002-88866 Supplier Medchemexpress LLC Supplier No. HY1614310MG
Cilengitide trifluoroacetate is the trifluoroacetate (TFA) salt of cilengitide, a cyclic RGD peptide that selectively inhibits αvβ3 and αvβ5 integrin receptors. It is provided as a high-purity research reagent for studies of integrin-mediated signaling, angiogenesis, and tumor biology.
  • Selective inhibitor of αvβ3 and αvβ5 integrins (IC50: 4 nM and 79 nM).
  • Supplied as the trifluoroacetate (TFA) salt form for research use.
  • High reported purity (~99.9%) suitable for laboratory studies.
  • Available in small research pack sizes (for example, 10 mg).