Learn More
Cayman Chemical CIlIobrevIn A 10mg
SDP
Catalog No. 502986870
Encompass_Preferred
A hedgehog pathway inhibitor that blocks Shh-induced hedgehog pathway activation (IC50 = 7 µM); inhibits primary cilia formation; inhibits dynein-dependent microtubial gliding and ATPase activities of the cytoplasmic AAA+ family; inhibits mTORC1 activity in uninfected and HCMV-infected cells
Catalog No. 50-298-6870 Supplier Cayman Chemical Supplier No. 2258710 MG
A hedgehog pathway inhibitor that blocks Shh-induced hedgehog pathway activation (IC50 = 7 µM); inhibits primary cilia formation; inhibits dynein-dependent microtubial gliding and ATPase activities of the cytoplasmic AAA+ family; inhibits mTORC1 activity in uninfected and HCMV-infected cells