Learn More
Medchemexpress LLC Ciliobrevin D | 1370554-01-0 | 99.4% | 10 MG
SDP
Catalog No. 5000233880
Encompass_Preferred
Ciliobrevin D is a cell-permeable, reversible inhibitor of the AAA+ ATPase motor cytoplasmic dynein, supplied as a purified solid for laboratory research. It is used to disrupt dynein-dependent processes such as Hedgehog signaling and primary cilia formation.
  • High purity suitable for research applications.
  • Molecular weight 392.62 g/mol; formula C17H8Cl3N3O2.
  • Light yellow to yellow powder for convenient handling.
  • 10 mg pack available for screening and assay development.
  • Stable when stored at recommended temperatures for extended periods.
Catalog No. 50-002-33880 Supplier Medchemexpress LLC Supplier No. HY12263210MG
Ciliobrevin D is a cell-permeable, reversible inhibitor of the AAA+ ATPase motor cytoplasmic dynein, supplied as a purified solid for laboratory research. It is used to disrupt dynein-dependent processes such as Hedgehog signaling and primary cilia formation.
  • High purity suitable for research applications.
  • Molecular weight 392.62 g/mol; formula C17H8Cl3N3O2.
  • Light yellow to yellow powder for convenient handling.
  • 10 mg pack available for screening and assay development.
  • Stable when stored at recommended temperatures for extended periods.