Learn More
Medchemexpress LLC Ciliobrevin D | 1370554-01-0 | 99.4% | 50 MG
SDP
Catalog No. 5000230635
Encompass_Preferred
Ciliobrevin D is a cell-permeable, reversible inhibitor of the AAA+ ATPase motor cytoplasmic dynein used in research to disrupt dynein-dependent microtubule gliding and ATPase activity, and to inhibit Hedgehog signaling and primary cilia formation.
  • High purity research-grade compound (99.4%).
  • Molecular formula C17H8Cl3N3O2 and molecular weight 392.62 g/mol.
  • Solid form, commonly supplied in small milligram quantities, including 50 mg.
  • Cell-permeable and reversible inhibitor of cytoplasmic dynein activity for in vitro studies.
  • Useful for studies of Hedgehog signaling, cilia formation, and microtubule dynamics.
Catalog No. 50-002-30635 Supplier Medchemexpress LLC Supplier No. HY12263250MG
Ciliobrevin D is a cell-permeable, reversible inhibitor of the AAA+ ATPase motor cytoplasmic dynein used in research to disrupt dynein-dependent microtubule gliding and ATPase activity, and to inhibit Hedgehog signaling and primary cilia formation.
  • High purity research-grade compound (99.4%).
  • Molecular formula C17H8Cl3N3O2 and molecular weight 392.62 g/mol.
  • Solid form, commonly supplied in small milligram quantities, including 50 mg.
  • Cell-permeable and reversible inhibitor of cytoplasmic dynein activity for in vitro studies.
  • Useful for studies of Hedgehog signaling, cilia formation, and microtubule dynamics.