Learn More
Selleck Chemical LLC Ciliobrevin D-5mg
SDP
Catalog No. 502389670
Encompass_Preferred
Ciliobrevin D (compound 5) is a cell-permeable, reversible and specific antagonist of AAA+ (ATPases associated with diverse cellular activities) ATPase motor cytoplasmic dynein. Ciliobrevin D perturbs primary cilia formation and blocks Hedgehog (Hh) signaling.
Catalog No. 50-238-9670 Supplier Selleck Chemical LLC Supplier No. S97435MG
Ciliobrevin D (compound 5) is a cell-permeable, reversible and specific antagonist of AAA+ (ATPases associated with diverse cellular activities) ATPase motor cytoplasmic dynein. Ciliobrevin D perturbs primary cilia formation and blocks Hedgehog (Hh) signaling.