Learn More
TARGETMOL CHEMICALS INC CILIOBREVIN D 5MG
SDP
Catalog No. 502581266
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ciliobrevin D is a AAA+ ATPase motor cytoplasmic dynein inhibitor. Ciliobrevin D inhibits Hedgehog (Hh) signaling and primary cilia formation and it also inhibits dynein-dependent microtubule gliding and ATPase activity in vitro. purity: 100%
Catalog No. 50-258-1266 Supplier TARGETMOL CHEMICALS INC Supplier No. T149655MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ciliobrevin D is a AAA+ ATPase motor cytoplasmic dynein inhibitor. Ciliobrevin D inhibits Hedgehog (Hh) signaling and primary cilia formation and it also inhibits dynein-dependent microtubule gliding and ATPase activity in vitro. purity: 100%