Learn More
Medchemexpress LLC HY-122632 5mg Medchemexpress, Ciliobrevin D CAS:1370554-01-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1226325MG
Encompass_Preferred
Medchemexpress, HY-122632 5mg Ciliobrevin D CAS:1370554-01-0 Ciliobrevin D is a cell-permeable, reversible and specific inhibitor of AAA+ ATPase motor cytoplasmic dynein. Ciliobrevin D inhibits Hedgehog (Hh) signaling and primary cilia formation. Ciliobrevin D inhibits dynein-dependent microtubule gliding and ATPase activity in vitro[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9930