Learn More
Selleck Chemical LLC Cilnidipine S1293-200mg
SDP
Catalog No. 503108345
Encompass_Preferred
Cilnidipine is a unique L-type and N-type calcium channel blocker used for high blood pressure treatment
Catalog No. 50-310-8345 Supplier Selleck Chemical LLC Supplier No. S1293200MG
Cilnidipine is a unique L-type and N-type calcium channel blocker used for high blood pressure treatment