Learn More
Selleck Chemical LLC Cilnidipine 50mg 132203-70-4 FRC-8653
SDP
Catalog No. 507488
Encompass_Preferred
Cilnidipine (FRC-8653) is a unique L-type and N-type calcium channel blocker, used for high blood pressure treatment. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-748-8 Supplier Selleck Chemical LLC Supplier No. S129350MG
Cilnidipine (FRC-8653) is a unique L-type and N-type calcium channel blocker, used for high blood pressure treatment. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.