Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cilnidipine, Tocris Bioscience™

Catalog No. 262910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-291-0 10 mg
26-295-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-291-0 Supplier R&D Systems Supplier No. 2629/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Ca2+ channel blocker (dual L/N-type)

Dual L- and N-type calcium channel blocker that displays antihypertensive, sympatholytic and neuroprotective activity. Lowers mean blood pressure and reduces the size of cerebral infarction in the rat model of focal brain ischemia.

Chemical Identifiers

CAS 132203-70-4
Molecular Formula C27H28N2O7
Molecular Weight (g/mol) 492.528
InChI Key KJEBULYHNRNJTE-DHZHZOJOSA-N
Synonym cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester
PubChem CID 5282138
ChEBI CHEBI:31399
IUPAC Name 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC

Specifications

Quantity 10 mg
Formula Weight 492.52
Percent Purity >99%
Chemical Name or Material Cilnidipine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.