Learn More
Medchemexpress LLC Cilobradine hydrochloride | 186097-54-1 | 99.7% | 519.07 | 1 ML
SDP
Catalog No. 5000385173
Encompass_Preferred
Cilobradine hydrochloride is a HCN channel blocker. It induces cardiac pacemaker currents and produces antidepressant effects, making it a valuable tool for research purposes.
  • Induces cardiac pacemaker currents.
  • Produces antidepressant effects.
  • Acts as a HCN channel blocker.
  • For research use only.
Catalog No. 50-003-85173 Supplier Medchemexpress LLC Supplier No. HY18940A10MM/1ML
Cilobradine hydrochloride is a HCN channel blocker. It induces cardiac pacemaker currents and produces antidepressant effects, making it a valuable tool for research purposes.
  • Induces cardiac pacemaker currents.
  • Produces antidepressant effects.
  • Acts as a HCN channel blocker.
  • For research use only.