Learn More
Medchemexpress LLC Cilobradine hydrochloride | 186097-54-1 | 99.7% | 519.07 | 50 MG
SDP
Catalog No. 5000385242
Encompass_Preferred
Cilobradine hydrochloride is a HCN channel blocker that induces cardiac pacemaker currents and produces antidepressant effects. It is intended for research use only.
  • Induces a use-dependent blockade of cardiac pacemaker current in rabbit sino-atrial cells.
  • Blocks HCN channel currents in HEK293 cells expressing human HCN1, HCN2, HCN3, and HCN4 channels in a dose- and treatment-dependent manner, and its steady-state inhibition of different HCN isoforms is not isoform-specific.
  • Can prolong the latency of the a-wave or b-wave of the rat electroretinogram (ERG).
  • Produces a dose-dependent waveform change after the b-wave at a specific light stimulus brightness.
  • Can reverse depressive-like behavior, and the effect lasts for about two weeks.
  • Increases time in social interaction and reduces time in the corner zone.
  • Increases sucrose preference and reduces immobility time in the forced swim test.
Catalog No. 50-003-85242 Supplier Medchemexpress LLC Supplier No. HY18940A50MG
Cilobradine hydrochloride is a HCN channel blocker that induces cardiac pacemaker currents and produces antidepressant effects. It is intended for research use only.
  • Induces a use-dependent blockade of cardiac pacemaker current in rabbit sino-atrial cells.
  • Blocks HCN channel currents in HEK293 cells expressing human HCN1, HCN2, HCN3, and HCN4 channels in a dose- and treatment-dependent manner, and its steady-state inhibition of different HCN isoforms is not isoform-specific.
  • Can prolong the latency of the a-wave or b-wave of the rat electroretinogram (ERG).
  • Produces a dose-dependent waveform change after the b-wave at a specific light stimulus brightness.
  • Can reverse depressive-like behavior, and the effect lasts for about two weeks.
  • Increases time in social interaction and reduces time in the corner zone.
  • Increases sucrose preference and reduces immobility time in the forced swim test.