Learn More
TARGETMOL CHEMICALS INC CILOMILAST 10MG
SDP
Catalog No. 502579358
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cilomilast (SB-207499) is a potent PDE4 inhibitor with IC50 of about 110 nM has anti-inflammatory activity and low central nervous system activity. Phase 3. purity: 99%
Catalog No. 50-257-9358 Supplier TARGETMOL CHEMICALS INC Supplier No. T6445
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cilomilast (SB-207499) is a potent PDE4 inhibitor with IC50 of about 110 nM has anti-inflammatory activity and low central nervous system activity. Phase 3. purity: 99%