Learn More
Cayman Chemical Cilostamide, Purity: ≥98%, CAS: 68550-75-4, 10 mg
SDP
Catalog No. 502971717
Encompass_Preferred
A selective inhibitor of PDE3A and PDE3B (IC50s = 27 and 50 nM, respectively); inhibits thrombin-induced platelet aggregation (IC50 = 1.1 μM)
Catalog No. 50-297-1717 Supplier Cayman Chemical Supplier No. 1445510 MG
A selective inhibitor of PDE3A and PDE3B (IC50s = 27 and 50 nM, respectively); inhibits thrombin-induced platelet aggregation (IC50 = 1.1 μM)