Learn More
Enzo Life Sciences Cilostamide (25mg). CAS: 68550-75-4
SDP
Supplier:  Enzo Life Sciences BMLPD1250025
Encompass_Preferred
Phosphodiesterase PDE3 inhibitor (IC50=5 nM). Blocks PKB/Akt signaling pathway by inhibition of Akt-activated PDE3B. Reverses the established effects of leptin on food intake and body weight. Blocks the leptin-induced tyrosine phosphorylation of STAT3 at the hypothalamic level. Alternative name: OCP 3689, OPS 3689, N-Cyclohexyl-N-methyl-4-(1,2-dihydro-2-oxo-6-quinolyloxy)butyramide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1373