Learn More
Medchemexpress LLC HY-101312 5mg Medchemexpress, Cilostamide CAS:68550-75-4 Purity:>98%
SDP
Catalog No. 501932093
Encompass_Preferred
Medchemexpress, HY-101312 5mg Cilostamide CAS:68550-75-4 Cilostamide is a selective and potent PDE3 inhibitor, with IC 50 s of 27 nM and 50 nM for PDE3A and PDE3B, respectively, and has antithrombotic and anti-intimal hyperplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2093 Supplier Medchemexpress LLC Supplier No. HY1013125MG
Medchemexpress, HY-101312 5mg Cilostamide CAS:68550-75-4 Cilostamide is a selective and potent PDE3 inhibitor, with IC 50 s of 27 nM and 50 nM for PDE3A and PDE3B, respectively, and has antithrombotic and anti-intimal hyperplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.