Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cilostamide

Catalog No. AAJ65319MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65319MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65319MA Supplier Thermo Scientific Chemicals Supplier No. J65319MA
  • $193.90
    $153.65 / Each of 1

    Save $40.25

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 68550-75-4 | C20H26N2O3 | 342.439 g/mol

A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68550-75-4
Molecular Formula C20H26N2O3
Molecular Weight (g/mol) 342.439
MDL Number MFCD00673958
InChI Key UIAYVIIHMORPSJ-UHFFFAOYSA-N
Synonym cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
PubChem CID 2753
IUPAC Name N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
SMILES CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3

Specifications

Color White
Quantity 10 mg
Formula Weight 342.43
Physical Form Powder
Chemical Name or Material Cilostamide
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.