Learn More
Enzo Life Sciences Cilostazol (50mg). CAS: 73963-72-1
SDP
Supplier:  Enzo Life Sciences BMLPD1270050
Encompass_Preferred
Cilostazol is a PDE3A inhibitor (0.2µM) and a unique antithrombotic agent. It inhibits cell death by activating maxi-K channels. Purity: ≥98%. Solubility: Soluble in DMSO (18mg/ml) or ethanol (4mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1375