Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cilostazol, 98%, Thermo Scientific Chemicals

Catalog No. AAJ62301MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62301MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62301MA Supplier Thermo Scientific Chemicals Supplier No. J62301MA
Add to Cart

missing translation for 'validMainframeMsg'

An inhibitor of phosphodiesterase III

An inhibitor of phosphodiesterase III

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73963-72-1
Molecular Formula C20H27N5O2
Molecular Weight (g/mol) 369.469
MDL Number MFCD00866780
InChI Key RRGUKTPIGVIEKM-UHFFFAOYSA-N
Synonym cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
PubChem CID 2754
ChEBI CHEBI:31401
IUPAC Name 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4

Specifications

Color White
Quantity 10 mg
Merck Index 14,2277
Formula Weight 369.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Cilostazol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber VC8277500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.