Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cilostazol 98.0+%, TCI America™
SDP

Catalog No. C25875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25875G 5 g
C25871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25875G Supplier TCI America Supplier No. C25875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73963-72-1
Molecular Formula C20H27N5O2
Molecular Weight (g/mol) 369.469
MDL Number MFCD00866780
InChI Key RRGUKTPIGVIEKM-UHFFFAOYSA-N
Synonym cilostazol, pletal, cilostazole, pletaal, cilostazolum, cilostazolum inn-latin, opc 21, opc-21, cilostazol inn:jan, pletal tn
PubChem CID 2754
ChEBI CHEBI:31401
IUPAC Name 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CCC(CC1)N2C(=NN=N2)CCCCOC3=CC4=C(C=C3)NC(=O)CC4

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 369.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cilostazol
RTECSNumber VC8277500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.