Learn More
Aviva Systems Biology Corp CILP Antibody - C-terminal region (ARP64683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CILP. It was validated on Western Blot. . Alternate names: CILP-1, HsT18872ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-335