Learn More
American Radiolabeled Chemicals Inc CIMETIDINE 14C 50UC
SDP
Catalog No. 50352921
Encompass_Preferred
Cimetidine-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 252.34, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-921 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 363850UC
Add to Cart
Add to Cart
Request a Quote
Cimetidine-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 252.34, Shipped in Dry Ice, Exclusive