Learn More
Medchemexpress LLC Cimetidine-d3 | 1185237-29-9 | C10H13D3N6S | 1 MG
SDP
Catalog No. 5000354347
Encompass_Preferred
Cimetidine-d3 is the deuterium-labeled form of Cimetidine. Cimetidine (SKF-92334) is an orally active, inverse histamine H2 receptor antagonist with a Ki of 0.6 μM. It functions as an inverse agonist and has demonstrated anti-cancer and anti-inflammatory activities.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-54347 Supplier Medchemexpress LLC Supplier No. HY14289S1MG
Cimetidine-d3 is the deuterium-labeled form of Cimetidine. Cimetidine (SKF-92334) is an orally active, inverse histamine H2 receptor antagonist with a Ki of 0.6 μM. It functions as an inverse agonist and has demonstrated anti-cancer and anti-inflammatory activities.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.