Learn More
Medchemexpress LLC Cimetidine (SKF-92334) | 51481-61-9 | 99.7% | C10H16N6S | 1 ML
SDP
Catalog No. 5000354206
Encompass_Preferred
Cimetidine (SKF-92334) is an orally active, inverse, and BBB-permeable histamine H2 receptor antagonist. It is primarily known as a gastric acid reducer and can be used for duodenal and gastric ulcers research. Additionally, Cimetidine demonstrates anti-cancer and anti-inflammatory activities.
  • Orally active histamine H2 receptor antagonist
  • Gastric acid reducer
  • Anti-cancer and anti-inflammatory activity
  • For research use only
Catalog No. 50-003-54206 Supplier Medchemexpress LLC Supplier No. HY1428910MM/1ML
Cimetidine (SKF-92334) is an orally active, inverse, and BBB-permeable histamine H2 receptor antagonist. It is primarily known as a gastric acid reducer and can be used for duodenal and gastric ulcers research. Additionally, Cimetidine demonstrates anti-cancer and anti-inflammatory activities.
  • Orally active histamine H2 receptor antagonist
  • Gastric acid reducer
  • Anti-cancer and anti-inflammatory activity
  • For research use only