Learn More
Medchemexpress LLC Cimetidine sulfoxide | 54237-72-8 | 97.0% | C10H16N6OS | 10MG
SDP
Catalog No. 5000261335
Encompass_Preferred
Cimetidine sulfoxide is the sulfoxide metabolite of cimetidine provided for research and analytical applications (CAS 54237-72-8). Supplied as a research-grade solid, it is used as a reference standard in studies of histamine H2-receptor antagonists, drug metabolism, and analytical method development.
  • High-purity reference standard suitable for analytical assays.
  • Useful for metabolic and pharmacokinetic studies.
  • Available in small milligram pack sizes for research use.
  • Documentation available: COA, SDS, and handling instructions.
  • Stable solid form for storage and long-term use.
Catalog No. 50-002-61335 Supplier Medchemexpress LLC Supplier No. HY13633810MG
Cimetidine sulfoxide is the sulfoxide metabolite of cimetidine provided for research and analytical applications (CAS 54237-72-8). Supplied as a research-grade solid, it is used as a reference standard in studies of histamine H2-receptor antagonists, drug metabolism, and analytical method development.
  • High-purity reference standard suitable for analytical assays.
  • Useful for metabolic and pharmacokinetic studies.
  • Available in small milligram pack sizes for research use.
  • Documentation available: COA, SDS, and handling instructions.
  • Stable solid form for storage and long-term use.