Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cimetidine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601705 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-705 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-705 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C149125KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C1491, 51481-61-9, C10H16N6S

Spectrum™ Chemical Cimetidine, USP is a histamine and H2 receptor antagonist used to treat peptic ulcers and heartburn. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 51481-61-9
CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 1%
Melting Range 139-144°C
Packaging Fiber Drum
Molecular Formula C10H16N6S
Quantity 25 kg
InChI Key AQIXAKUUQRKLND-UHFFFAOYSA-N
SMILES CN=C(NCCSCC1=C(C)NC=N1)NC#N
IUPAC Name N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
Molecular Weight (g/mol) 252.34
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.