Learn More
Medchemexpress LLC Cimetropium Bromide | 51598-60-8 | 438.36 | 10 MG
SDP
Catalog No. 5000448568
Encompass_Preferred
Cimetropium Bromide (DA-3177) is a muscarinic acetylcholine receptor (mAChR) antagonist used for the long-term treatment of irritable bowel syndrome. It is a competitive antagonist of muscarinic-mediated contractions in isolated colonic preparations. In vivo, it acts as a potent inhibitor of large bowel motility.
  • Used for long-term treatment of irritable bowel syndrome
  • Competitive antagonist of muscarinic-mediated contractions
  • Potent antimuscarinic effect in inhibiting longitudinal muscle contraction
  • Potent inhibitor of large bowel motility evoked by exogenous and endogenous stimuli
Catalog No. 50-004-48568 Supplier Medchemexpress LLC Supplier No. HYU0010610MG
Cimetropium Bromide (DA-3177) is a muscarinic acetylcholine receptor (mAChR) antagonist used for the long-term treatment of irritable bowel syndrome. It is a competitive antagonist of muscarinic-mediated contractions in isolated colonic preparations. In vivo, it acts as a potent inhibitor of large bowel motility.
  • Used for long-term treatment of irritable bowel syndrome
  • Competitive antagonist of muscarinic-mediated contractions
  • Potent antimuscarinic effect in inhibiting longitudinal muscle contraction
  • Potent inhibitor of large bowel motility evoked by exogenous and endogenous stimuli