Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cimiracemoside C | 256925-92-5 | 10 MG
SDP

Catalog No. 5000417211
Encompass_Preferred

Cimiracemoside C is an active component of Cimicifuga racemosa. This compound activates AMPK and shows potential activity against diabetes.

  • Active component of Cimicifuga racemosa
  • Activates AMPK
  • Potential activity against diabetes
  • High purity: 99.58%
  • Molecular weight: 620.81
  • Molecular formula: C35H56O9
  • Appearance: White to off-white solid
  • Store at 4°C, protected from light
  • Soluble in DMSO (5 mg/mL)
  • Insoluble in water

Catalog No. 50-004-17211 Supplier Medchemexpress LLC Supplier No. HYN697110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cimiracemoside C is an active component of Cimicifuga racemosa. This compound activates AMPK and shows potential activity against diabetes.

  • Active component of Cimicifuga racemosa
  • Activates AMPK
  • Potential activity against diabetes
  • High purity: 99.58%
  • Molecular weight: 620.81
  • Molecular formula: C35H56O9
  • Appearance: White to off-white solid
  • Store at 4°C, protected from light
  • Soluble in DMSO (5 mg/mL)
  • Insoluble in water

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.