Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cimiracemoside D | 290821-39-5 | MFCD28411517 | 678.85 | 1 ML
SDP

Catalog No. 5000416889
Encompass_Preferred

Cimiracemoside D is a natural product isolated from Actaea racemosa, appearing as a white to off-white solid. It exhibits solubility in DMSO (100 mg/mL, 147.31 mM with sonication) and detailed protocols for preparing stock solutions and in vivo dissolution are available.

  • Purity of 99.89%
  • Isolated from Actaea racemosa
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL, 147.31 mM)
  • Includes protocols for stock solution preparation
  • Includes protocols for in vivo dissolution using various solvents
  • Powder storage at -20°C for 3 years, 4°C for 2 years
  • In-solvent storage at -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-16889 Supplier Medchemexpress LLC Supplier No. HYN090010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cimiracemoside D is a natural product isolated from Actaea racemosa, appearing as a white to off-white solid. It exhibits solubility in DMSO (100 mg/mL, 147.31 mM with sonication) and detailed protocols for preparing stock solutions and in vivo dissolution are available.

  • Purity of 99.89%
  • Isolated from Actaea racemosa
  • Appears as a white to off-white solid
  • Soluble in DMSO (100 mg/mL, 147.31 mM)
  • Includes protocols for stock solution preparation
  • Includes protocols for in vivo dissolution using various solvents
  • Powder storage at -20°C for 3 years, 4°C for 2 years
  • In-solvent storage at -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.