Learn More
Medchemexpress LLC HY-118643 10mg Medchemexpress, Cimlanod CAS:1620330-72-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11864310MG
Encompass_Preferred
Medchemexpress, HY-118643 10mg Cimlanod CAS:1620330-72-4 Cimlanod (BMS-986231) is a second-generation Nitroxyl (HNO) donor for heart failure. Cimlanod (BMS-986231) delivers HNO via pH-dependent chemical breakdown when exposed to the neutral pH environment of the bloodstream. Cimlanod (BMS-986231) possesses positive lusitropic and inotropic as well as vasodilatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0919