Learn More
Bia Separartions Inc Oligo dT18 (C12 Linker) - Affinity, 1 mL (2 um)
Catalog No. NC1870109
Encompass
CIMmultus Oligo dT18 (C12) is a monolithic column with 2 μm channel size and a column volume of 1 mL. The column is intended for initial screening, method development, and lab scale purifications.
Catalog No. NC1870109 Supplier Bia Separartions Inc Supplier No. 311.12192
CIMmultus Oligo dT18 (C12) is a monolithic column with 2 μm channel size and a column volume of 1 mL. The column is intended for initial screening, method development, and lab scale purifications.