Learn More
MSC CIMTECH 500 5 GAL PAIL
Catalog No. NC2205070
Encompass
Cimcool CIMTECH 500 is a versatile cutting and grinding fluid designed for various machining operations. It provides excellent performance for applications involving carbon steel, cast iron, ductile iron, and stainless steel, and is supplied in a 5 gal pail.
  • Excellent foam control even in turbulent grinding systems
  • Good lubricity for extended tool and wheel life
  • Suitable for drilling, reaming, milling, and boring
This item is not returnable. View return policy
Catalog No. NC2205070 Supplier MSC Supplier No. 05930136
Cimcool CIMTECH 500 is a versatile cutting and grinding fluid designed for various machining operations. It provides excellent performance for applications involving carbon steel, cast iron, ductile iron, and stainless steel, and is supplied in a 5 gal pail.
  • Excellent foam control even in turbulent grinding systems
  • Good lubricity for extended tool and wheel life
  • Suitable for drilling, reaming, milling, and boring