Learn More
HATCHING TIME LLC Cimuka Egg Incubator Setter/Hatcher, 3200 Chicken Egg Capacity, Automatic Temperature & Humidity
Catalog No. NC3995687
Encompass
  • This “All in / all out” egg incubator setter/hatcher for commercial use provides fully automated incubation with automatic egg turners and humidity/temperature sensors for high hatching rates and energy efficiency. It features two trolleys, an insulated cabinet, and an advanced control panel with monitoring and alarm functions.
  • All in / all out design with two trolleys for easy setting and hatching.
  • Fully automated incubation with egg turners, temperature, and humidity sensors.
  • Advanced control panel provides monitoring and alarm functions.
  • Energy efficient with durable ABS plastic walls for cleaning.
  • Active periodic cooling enhances natural processes for increased hatch rates.
  • IP55 certified ventilation ensures consistent conditions.
  • Humisonic™ system offers precise humidity regulation.
Catalog No. NC3995687 Supplier HATCHING TIME LLC Supplier No. T3200SHT
  • This “All in / all out” egg incubator setter/hatcher for commercial use provides fully automated incubation with automatic egg turners and humidity/temperature sensors for high hatching rates and energy efficiency. It features two trolleys, an insulated cabinet, and an advanced control panel with monitoring and alarm functions.
  • All in / all out design with two trolleys for easy setting and hatching.
  • Fully automated incubation with egg turners, temperature, and humidity sensors.
  • Advanced control panel provides monitoring and alarm functions.
  • Energy efficient with durable ABS plastic walls for cleaning.
  • Active periodic cooling enhances natural processes for increased hatch rates.
  • IP55 certified ventilation ensures consistent conditions.
  • Humisonic™ system offers precise humidity regulation.