Learn More
Cayman Chemical Cinnabarinic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502919381
Encompass_Preferred
An internal standard for the quantification of cinnabarinic acid by GC- or LC-MS
Catalog No. 50-291-9381 Supplier Cayman Chemical Supplier No. 372091 MG
An internal standard for the quantification of cinnabarinic acid by GC- or LC-MS