Learn More
Medchemexpress LLC HY-70037 10mg Medchemexpress, Cinacalcet CAS:226256-56-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7003710MG
Encompass_Preferred
Medchemexpress, HY-70037 10mg Cinacalcet CAS:226256-56-0 Cinacalcet (AMG 073) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0962