Learn More
Medchemexpress LLC Cinacalcet-d3 hydrochloride | 364782-34-3 | 97.6% | C22H20D3ClF3N | 500 UG
SDP
Catalog No. 5000434242
Encompass_Preferred
Cinacalcet-d3 (hydrochloride) is the deuterium labeled Cinacalcet (hydrochloride). It is an orally active, allosteric agonist of the Ca receptor (CaR), used for cardiovascular disease treatment. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compound.
  • Endogenous metabolite.
  • Inhibitor.
  • Targets CaSR and is relevant for cardiovascular disease.
Catalog No. 50-004-34242 Supplier Medchemexpress LLC Supplier No. HY70037AS500UG
Cinacalcet-d3 (hydrochloride) is the deuterium labeled Cinacalcet (hydrochloride). It is an orally active, allosteric agonist of the Ca receptor (CaR), used for cardiovascular disease treatment. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compound.
  • Endogenous metabolite.
  • Inhibitor.
  • Targets CaSR and is relevant for cardiovascular disease.