Learn More
Medchemexpress LLC Cinacalcet-d4 hydrochloride | 2514927-75-2 | ≥98.0% | 397.90 g/mol | C22H19D4ClF3N | 2.5 MG
SDP
Catalog No. 5000284441
Encompass_Preferred
Cinacalcet-d4 hydrochloride is a deuterium-labeled analogue of cinacalcet provided as the hydrochloride salt for use as an isotopically labeled internal standard in analytical and research workflows, particularly LC-MS quantitation. It has molecular formula C22H19D4ClF3N, molecular weight 397.90 g/mol, and typical purity ≥98.0%.
  • Deuterium-labeled internal standard suitable for LC-MS quantitation.
  • Hydrochloride salt form for stable handling and solubility in common solvents.
  • D4 isotopic labeling reduces interference from native analyte peaks.
  • High purity (≥98.0%) supports accurate and reproducible results.
  • Available in small mass packages for reference standard and trace-level analysis.
Catalog No. 50-002-84441 Supplier Medchemexpress LLC Supplier No. HYW6539782.5MG
Cinacalcet-d4 hydrochloride is a deuterium-labeled analogue of cinacalcet provided as the hydrochloride salt for use as an isotopically labeled internal standard in analytical and research workflows, particularly LC-MS quantitation. It has molecular formula C22H19D4ClF3N, molecular weight 397.90 g/mol, and typical purity ≥98.0%.
  • Deuterium-labeled internal standard suitable for LC-MS quantitation.
  • Hydrochloride salt form for stable handling and solubility in common solvents.
  • D4 isotopic labeling reduces interference from native analyte peaks.
  • High purity (≥98.0%) supports accurate and reproducible results.
  • Available in small mass packages for reference standard and trace-level analysis.