Learn More
Medchemexpress LLC HY-70037A 10mg Medchemexpress, Cinacalcet (hydrochloride) CAS:364782-34-3 Purity:>98%
SDP
Catalog No. 502030963
Encompass_Preferred
Medchemexpress, HY-70037A 10mg Cinacalcet (hydrochloride) CAS:364782-34-3 Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0963 Supplier Medchemexpress LLC Supplier No. HY70037A10MG
Medchemexpress, HY-70037A 10mg Cinacalcet (hydrochloride) CAS:364782-34-3 Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.