Learn More
Medchemexpress LLC Cinacalcet (hydrochloride) | 364782-34-3 | 100.0% | C22H23ClF3N | 100 MG
SDP
Catalog No. 5000434706
Encompass_Preferred
Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of the calcium-sensing receptor (CaR), primarily utilized for the treatment of cardiovascular diseases. This high-purity compound is provided as a white to off-white solid, suitable for various research applications.
  • Orally active, allosteric agonist of Ca receptor
  • Used in cardiovascular disease treatment
  • High purity of 100.0%
  • Good solubility in various solvents
  • Comprehensive documentation available
  • For research use only
Catalog No. 50-004-34706 Supplier Medchemexpress LLC Supplier No. HY70037A100MG
Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of the calcium-sensing receptor (CaR), primarily utilized for the treatment of cardiovascular diseases. This high-purity compound is provided as a white to off-white solid, suitable for various research applications.
  • Orally active, allosteric agonist of Ca receptor
  • Used in cardiovascular disease treatment
  • High purity of 100.0%
  • Good solubility in various solvents
  • Comprehensive documentation available
  • For research use only