Learn More
Medchemexpress LLC Cinacalcet (hydrochloride) | 364782-34-3 | 99.9% | C22H23ClF3N | 1 ML
SDP
Catalog No. 5000434307
Encompass_Preferred
Cinacalcet hydrochloride is an orally active, allosteric agonist of the Ca receptor (CaR), used for cardiovascular disease treatment. It is intended for identified uses as laboratory chemicals and in the manufacture of substances.
  • Orally active allosteric agonist
  • Used for cardiovascular disease treatment
  • For research use only
  • Purity of 99.88%
  • Solid form with melting point of 182.0-182.3°C
Catalog No. 50-004-34307 Supplier Medchemexpress LLC Supplier No. HY70037A10MM/1ML
Cinacalcet hydrochloride is an orally active, allosteric agonist of the Ca receptor (CaR), used for cardiovascular disease treatment. It is intended for identified uses as laboratory chemicals and in the manufacture of substances.
  • Orally active allosteric agonist
  • Used for cardiovascular disease treatment
  • For research use only
  • Purity of 99.88%
  • Solid form with melting point of 182.0-182.3°C