Learn More
Cayman Chemical Cinacalcet (hydrochloride), Purity: ≥95%, CAS: 364782-34-3, 100 mg
SDP
Catalog No. 502969210
Encompass_Preferred
An allosteric agonist of the CaSR (EC50 = 79.4 nM in HEK293T cells expressing the human receptor); decreases plasma levels of PTH in rats from 0.1-10 mg/kg; decreases plasma levels of PTH and parathyroid cell proliferation in a mouse model of primary hyperparathyroidism
Catalog No. 50-296-9210 Supplier Cayman Chemical Supplier No. 16042100 MG
An allosteric agonist of the CaSR (EC50 = 79.4 nM in HEK293T cells expressing the human receptor); decreases plasma levels of PTH in rats from 0.1-10 mg/kg; decreases plasma levels of PTH and parathyroid cell proliferation in a mouse model of primary hyperparathyroidism