Learn More
Medchemexpress LLC HY-14181 5mg Medchemexpress, Cinaciguat CAS:329773-35-5 Purity:>98%
SDP
Catalog No. 502081571
Encompass_Preferred
Medchemexpress, HY-14181 5mg Cinaciguat CAS:329773-35-5 Cinaciguat is an activator of guanylate cyclase (sGC), and used for acute decompensated heart failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1571 Supplier Medchemexpress LLC Supplier No. HY141815MG
Medchemexpress, HY-14181 5mg Cinaciguat CAS:329773-35-5 Cinaciguat is an activator of guanylate cyclase (sGC), and used for acute decompensated heart failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.