Learn More
Medchemexpress LLC HY-14181A 2mg Medchemexpress, Cinaciguat (hydrochloride) CAS:646995-35-9 Purity:>98%
SDP
Catalog No. 501932697
Encompass_Preferred
Medchemexpress, HY-14181A 2mg Cinaciguat (hydrochloride) CAS:646995-35-9 Cinaciguat hydrochloride is a potent soluble guanylate cyclase ( GC ) activator with EC 50 of 15 nM in platelets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2697 Supplier Medchemexpress LLC Supplier No. HY14181A2MG
Medchemexpress, HY-14181A 2mg Cinaciguat (hydrochloride) CAS:646995-35-9 Cinaciguat hydrochloride is a potent soluble guanylate cyclase ( GC ) activator with EC 50 of 15 nM in platelets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.