Learn More
Medchemexpress LLC Cinaciguat (hydrochloride) | 646995-35-9 | 98.90% | C36H40ClNO5 | 10 MM * 1 ML
SDP
Catalog No. 5000353757
Encompass_Preferred
Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator with an EC50 of 15 nM in platelets. It is available as a 10 mM * 1 mL solution in DMSO and is suitable for various laboratory applications including research into cardiovascular disease and related classifications.
  • Potent soluble guanylate cyclase (GC) activator
  • EC50 of 15 nM in platelets
  • Activates apo-sGC
  • Modulates vasorelaxation and blood pressure
Catalog No. 50-003-53757 Supplier Medchemexpress LLC Supplier No. HY14181A10MM/1ML
Add to Cart
Add to Cart
Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator with an EC50 of 15 nM in platelets. It is available as a 10 mM * 1 mL solution in DMSO and is suitable for various laboratory applications including research into cardiovascular disease and related classifications.
  • Potent soluble guanylate cyclase (GC) activator
  • EC50 of 15 nM in platelets
  • Activates apo-sGC
  • Modulates vasorelaxation and blood pressure