Learn More
Cayman Chemical Cinaciguat (hydrochloride), Purity: ≥98%, CAS: 646995-35-9, 10 mg
SDP
Catalog No. 502968173
Encompass_Preferred
An activator of sGC that binds to a regulatory site, resulting in activation in an NO-independent manner (Kd = 3.2 nM); reduces hypertension, limits cardiomyocyte hypertrophy, protects against ischemia/reperfusion injury, and reduces morbidity and mortality in response to endotoxic shock
Catalog No. 50-296-8173 Supplier Cayman Chemical Supplier No. 1746810 MG
An activator of sGC that binds to a regulatory site, resulting in activation in an NO-independent manner (Kd = 3.2 nM); reduces hypertension, limits cardiomyocyte hypertrophy, protects against ischemia/reperfusion injury, and reduces morbidity and mortality in response to endotoxic shock