Learn More
Cayman Chemical CInamoylglycIn 10mg
SDP
Catalog No. 502912052
Encompass_Preferred
A metabolite of trans-cinnamic acid; urinary levels are decreased in a mouse model of piperacillin- and tazobactam-induced gut microbiota colonization resistance; serum levels are increased in patients with CKD
Catalog No. 50-291-2052 Supplier Cayman Chemical Supplier No. 3637810 MG
A metabolite of trans-cinnamic acid; urinary levels are decreased in a mouse model of piperacillin- and tazobactam-induced gut microbiota colonization resistance; serum levels are increased in patients with CKD