Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cinanserin hydrochloride | 54-84-2 | MFCD00055609 | 99.5% | 376.94 g/mol | C20H25ClN2OS | 5 MG
SDP

Catalog No. 5000211521
Encompass_Preferred

Cinanserin hydrochloride is a research-grade serotonin (5-HT2) receptor antagonist supplied as the hydrochloride salt for use in in vitro and preclinical pharmacology studies. It is provided as a solid suitable for assay development, receptor-binding studies, and biochemical experiments.

  • Research-grade 5-HT2 receptor antagonist.
  • Supplied as the hydrochloride salt in solid form.
  • High purity (99.5%).
  • CAS number 54-84-2.
  • Molecular weight 376.94 g/mol.
  • Intended for in vitro and preclinical research use.
  • Store at 4 °C and protect from moisture.

Catalog No. 50-002-11521 Supplier Medchemexpress LLC Supplier No. HY1009435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cinanserin hydrochloride is a research-grade serotonin (5-HT2) receptor antagonist supplied as the hydrochloride salt for use in in vitro and preclinical pharmacology studies. It is provided as a solid suitable for assay development, receptor-binding studies, and biochemical experiments.

  • Research-grade 5-HT2 receptor antagonist.
  • Supplied as the hydrochloride salt in solid form.
  • High purity (99.5%).
  • CAS number 54-84-2.
  • Molecular weight 376.94 g/mol.
  • Intended for in vitro and preclinical research use.
  • Store at 4 °C and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.