Learn More
TARGETMOL CHEMICALS INC CINANSERIN HYDROCHLORIDE 5MG
SDP
Catalog No. 502581265
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cinanserin hydrochloride (SQ 10643) is a high affinity antagonist of the 5-HT2 receptor (Ki 41 nM) and a 3C-like protease inhibitor of severe acute respiratory syndrome coronavirus. purity: 99%
Catalog No. 50-258-1265 Supplier TARGETMOL CHEMICALS INC Supplier No. T149685MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cinanserin hydrochloride (SQ 10643) is a high affinity antagonist of the 5-HT2 receptor (Ki 41 nM) and a 3C-like protease inhibitor of severe acute respiratory syndrome coronavirus. purity: 99%