Learn More
MSC Bulk Strap Cinch Strap, Polypropylene and Zinc, 1 in. x 48 in., 1 ea.
Catalog No. 5000516756
Encompass
This cinch strap is designed for securing cargo and equipment, featuring a durable polypropylene webbing construction with a zinc cam buckle. The adjustable design provides a reliable hold for various applications requiring non-load-rated fastening.
  • Webbing width of 1 in.
  • Overall length of 48 in.
  • Constructed from polypropylene and zinc materials
  • Black color finish
  • Includes an adjustable cam buckle for secure tightening
  • Manufactured in the United States
Catalog No. 50-005-16756 Supplier MSC Supplier No. 13825971
This cinch strap is designed for securing cargo and equipment, featuring a durable polypropylene webbing construction with a zinc cam buckle. The adjustable design provides a reliable hold for various applications requiring non-load-rated fastening.
  • Webbing width of 1 in.
  • Overall length of 48 in.
  • Constructed from polypropylene and zinc materials
  • Black color finish
  • Includes an adjustable cam buckle for secure tightening
  • Manufactured in the United States