Learn More
AdipoGen Cinchocaine (5 g)
SDP
Supplier:  AdipoGen CDXC0694G005
Encompass_Preferred
Chemical. CAS: 85-79-0. Formula: C20H29N3O2. MW: 343.46. Synthetic. Cinchocaine is an amide local anesthetic. It is among the most potent and toxic of the long-acting local anesthetics, by blocking sodium channels. Shown to be a calpain activator inducing platelet apoptosis and to induce apoptosis in leukemia cells. This compound can be used as a reference compound.
Catalog No. 50-205-4347